Farnesol (mixture of isomers), Sigma-Aldrich®
Supplier: Merck
Synonyms: Farnesol, 3,7,11-Trimethyldodeca-2,6,10-trien-1-ol, TRANS,TRANS-3,7,11-TRIMETHYL-2,6,10-DODECATRIEN-1-OL, 2,6,10-Dodecatrien-1-ol, 3,7,11-trimethyl-, (2Z,6Z)-
MQ300
W247804-100G-K W247804-1KG-K
SIALW247804-100G-KEA 121 EUR
SIALW247804-100G-K SIALW247804-1KG-K
Farnesol (mixture of isomers), Sigma-Aldrich®
Farnesol (mixture of isomers)
Formula: C₁₅H₂₆O
MW: 222.37 g/mol
MDL Number: MFCD00002918
CAS Number: 4602-84-0
EINECS: 225-004-1