α-Chloro-o-xylene
Supplier: Thermo Scientific
Synonyms: 2-Methylbenzyl chloride
SB00681.1 SB00681.10 SB00681.50
MAYBSB00681.1EA 0 EUR
MAYBSB00681.1 MAYBSB00681.10 MAYBSB00681.50
α-Chloro-o-xylene
α-Chloro-o-xylene
Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 199
Melting Pt: –2 °C
Density: 1.08 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000899
CAS Number: 552-45-4
EINECS: 209-013-8
UN: 3265
ADR: 8 II
Merck Index: 12,10224