Benzofuran

Supplier: Thermo Scientific

Synonyms: 2,3-Benzofuran, Benzo[b]furan, Benzo(b)furan, 1-Benzofuran, Coumarone

SB00762.1 SB00762.10 SB00762.50
MAYBSB00762.1EA 0 EUR
MAYBSB00762.1 MAYBSB00762.10 MAYBSB00762.50
Benzofuran
Benzofuran
Formula: C₈H₆O
MW: 118.14 g/mol
Boiling Pt: 173
Melting Pt: < –18 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 50
MDL Number: MFCD00005847
CAS Number: 271-89-6
EINECS: 205-982-6
Merck Index: 13,01089

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR