3,5-Xylidine 97+%
Supplier: Thermo Fisher Scientific
Danger
160191000 160195000 160190050
ACRO160191000EA 40.9 EUR
ACRO160191000 ACRO160195000 ACRO160190050
3,5-Xylidine 97+%
3,5-Xylidine
Formula: C₈H₁₁N
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 9.8 °C
Density: 0.97 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00007813
CAS Number: 108-69-0
EINECS: 203-607-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217
Order Now
Specification Test Results
Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5560 to 1.5600 (20°C, 589 nm)
GC ≥97.0 %
Titration with HClO4 97.0 to 103.0 %