2,4-Di-tert-butylphenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1,3-Di(dimethylethyl)-4-hydroxybenzene
M-Clarity™ quality level = MQ100
137731-100G 137731-500G 137731-5G
SIAL137731-100GEA 30.3 EUR
SIAL137731-100G SIAL137731-500G SIAL137731-5G
2,4-Di-tert-butylphenol, Sigma-Aldrich®
2,4-Di-tert-butylphenol
Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 53…56 °C
Density: 0.887 g/cm³ (20 °C)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00008828
CAS Number: 96-76-4
EINECS: 202-532-0
UN: 2430
ADR: 8,III