o-Xylene ≥96% for HPLC
Supplier: Thermo Scientific
Warning
22902.M1EA 397 EUR
22902.M1 22902.K2 22902.K7
o-Xylene ≥96% for HPLC
o-Xylene
Purity: 96% min
CAS No.: 95-47-6
Molecular Formula: C8H10
Formula Weight: 106.17
Flash Point: 32°(89°F)
Physical Form: liquid
Density: 0.877
Refractive Index: 1.5050
Appearance: Colourless
Melting Point: -25°
Boiling Point: 143-145°
Beilstein Reference: 1815558
Merck Reference: 14,10081
UN No.: 1307
MDL No.: MFCD00008519
Formula: C₈H₁₀
MW: 106,17 g/mol
Boiling Pt: 143…145 °C
Melting Pt: –25 °C
Density: 0,877
Flash Pt: 32 °C (89 °F)
MDL Number: MFCD00008519
UN: 1307
ADR: 3,III
Merck Index: 14,10081
https://si.vwr.com/stibo/web/std.lang.all/85/44/768544.jpg