o-Xylene
Supplier: EHRENSTORFER GMBH
EHERC17945000EA
65.8
EUR
EHERC17945000
o-Xylene
o-Xylene
Formula:
C₈H₁₀ MW: 106,17 g/mol Boiling Pt: 144 °C (1013 hPa) Melting Pt: –25 °C Density: 0,88 g/cm³ (20 °C) Flash Pt: 30 °C (closed cup) |
MDL Number:
MFCD00008519 UN: 1307 ADR: 3,III Merck Index: 12,10214 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...