2,6-Di-tert-butyl-p-cresol
Supplier: Cayman Chemical
Synonyms: Butylated hydroxytoluene, Butylhydroxytoluene, 2,6-Di-tert-butyl-4-methylphenol, BHT
89910-500 89910-1
CAYM89910-500EA 24 EUR
CAYM89910-500 CAYM89910-1
2,6-Di-tert-butyl-p-cresol
2,6-Di-tert-butyl-p-cresol
Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 69…70 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00011644
CAS Number: 128-37-0
EINECS: 204-881-4
Merck Index: 13,01547