2,6-Di-tert-butyl-p-cresol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
34750-250G-F 34750-1KG-F
SIAL34750-250G-FEA 25.7 EUR
SIAL34750-250G-F SIAL34750-1KG-F
2,6-Di-tert-butyl-p-cresol, Sigma-Aldrich®
2,6-Di-tert-butyl-p-cresol
Formula: C₁₅H₂₄O
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 69…70 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00011644
CAS Number: 128-37-0
EINECS: 204-881-4
Merck Index: 13,01547