2,4,6-Tri-tert-butylphenol 97%
Supplier: Thermo Scientific
Danger
139330050 139330500 139335000
ACRO139330050EA 24.3 EUR
ACRO139330050 ACRO139330500 ACRO139335000
2,4,6-Tri-tert-butylphenol 97%
2,4,6-Tri-tert-butylphenol
Formula: C₁₈H₃₀O
MW: 262.44 g/mol
Boiling Pt: 277 °C (760 mmHg)
Melting Pt: 129…132 °C
Density: 0,919 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008821
CAS Number: 732-26-3
EINECS: 211-989-5
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Yellow to light brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
GC ≥96.0 %