1,2,4-Trimethylbenzene 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Pseudocumene
140090010 140090025 140090100
ACRO140090010EA 78.1 EUR
ACRO140090010 ACRO140090025 ACRO140090100
1,2,4-Trimethylbenzene 98%
1,2,4-Trimethylbenzene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 168 °C (760 mmHg)
Melting Pt: –44 °C
Density: 0.88 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00008527
CAS Number: 95-63-6
EINECS: 202-436-9
UN: 1993
ADR: 3 III
Merck Index: 12,08100
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5030 to 1.5050 (20°C, 589 nm)
GC ≥97.5 %