1,3,5-Triethylbenzene, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

132071-5G 132071-25G
SIAL132071-5GEA 72.8 EUR
SIAL132071-5G SIAL132071-25G
1,3,5-Triethylbenzene, Sigma-Aldrich®
1,3,5-Triethylbenzene
Formula: C₁₂H₁₈
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: 3 °C
Density: 0.8631 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009261
CAS Number: 102-25-0
EINECS: 203-017-3
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR