Pentamethylbenzene ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 1,2,3,4,5-Pentamethylbenzene

L04163.06 L04163.22 L04163.14
L04163.06EA 37.1 EUR
L04163.06 L04163.22 L04163.14
Pentamethylbenzene ≥99%
Pentamethylbenzene

Purity: 99% CAS No.: 700-12-9 Molecular Formula: C11H16 Formula Weight: 148.25 Flash Point: 91°(195°F) Melting Point: 50-51° Boiling Point: 231-232° Beilstein Reference: 1905349 UN No.: 1325 MDL No.: MFCD00008522

Formula: C₁₁H₁₆
MW: 148.25 g/mol
Boiling Pt: 231…232 °C
Melting Pt: 50…51 °C
Density: 0.917
Flash Pt: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008522
CAS Number: 700-12-9
EINECS: 211-837-8
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR