4-Phenylbenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Biphenyl-4-carbaldehyde, [1,1'-Biphenyl]-4-carbaldehyde, 4-Biphenylcarboxaldehyde, Biphenyl-4-carboxaldehyde, p-Biphenylylaldehyde, 4-Biphenylylaldehyde, 4-Biphenylcarbaldehyde
A15839.06 A15839.14 A15839.22
A15839.06EA 32.4 EUR
A15839.06 A15839.14 A15839.22
4-Phenylbenzaldehyde ≥98%
4-Phenylbenzaldehyde
Purity: 98+% CAS No.: 3218-36-8 Molecular Formula: C13H10O Formula Weight: 182.22 Flash Point: 172°(341°F) Melting Point: 57-61° Boiling Point: 184°/11mm Beilstein Reference: 606693 MDL No.: MFCD00006947
Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 184 °C (11 mmHg)
Melting Pt: 57…61 °C
Density: 1.107
Flash Pt: 172 °C (341 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006947
CAS Number: 3218-36-8
EINECS: 221-742-3