Benzyl benzoate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL12392-1L-FEA 183 EUR
SIAL12392-1L-F
Benzyl benzoate, Sigma-Aldrich®
Benzyl benzoate
Formula: C₁₄H₁₂O₂
MW: 212,25 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 17…20 °C
Density: 1,118 g/cm³ (20 °C)
Flash Pt: 158 °C (closed cup)
MDL Number: MFCD00003075
UN: 3082
ADR: 9,III
Merck Index: 13,01129

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR