N-Benzylaniline 99%
Supplier: Thermo Scientific
Synonyms: N-Phenylbenzylamine, ((Phenylmethyl)amino)benzene
417390250 417391000 417390050
ACRO417390250EA 52.6 EUR
ACRO417390250 ACRO417391000 ACRO417390050
N-Benzylaniline 99%
N-Benzylaniline
Formula: C₆H₅CH₂NHC₆H₅
MW: 183.25 g/mol
Boiling Pt: 306…307 °C (760 mmHg)
Melting Pt: 34…38 °C
Density: 1.06 g/cm³
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00003018
CAS Number: 103-32-2
EINECS: 203-100-4
Merck Index: 12,01161
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cee0f9ae18e00188e4a272cc12dbca2071f87d96&vid=22e30683618c7daab6a3a3013140064b6131ccea&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to yellow to light beige Crystalline powder or low melting mass
Infrared spectrum Conforms
Melting point 34 to 38 °C
GC ≥98.5 %
Titration with HClO4 ≥98.5 %