4,7-Dichloroquinoline
Supplier: Thermo Scientific
Warning
SB01458.1 SB01458.10 SB01458.50
MAYBSB01458.1EA 0 EUR
MAYBSB01458.1 MAYBSB01458.10 MAYBSB01458.50
4,7-Dichloroquinoline
4,7-Dichloroquinoline
Formula: C₉H₅Cl₂N
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00006774
CAS Number: 86-98-6
EINECS: 201-714-7