2-Quinolinol 98%
Supplier: Thermo Scientific
Danger
Synonyms: Quinolin-2(1H)-one, Quinolin-2-ol, Carbostyril, 2-Quinolone, 2-Hydroxyquinoline
418810100 418810010
ACRO418810100EA 238 EUR
ACRO418810100 ACRO418810010
2-Quinolinol 98%
2-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 197…200 °C
Density: 1,37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006743
CAS Number: 59-31-4
EINECS: 200-420-6
Merck Index: 13,01835
Order Now
Specification Test Results
Appearance White to brown-purple to purple Powder
Infrared spectrum Conforms
Melting point 194 to 200 °C
HPLC ≥97.5 %