7-Quinolinol 99%
Supplier: Thermo Scientific
Warning
Synonyms: Quinolin-7-ol, 7-Hydroxyquinoline
418820010 418820050
ACRO418820010EA 131 EUR
ACRO418820010 ACRO418820050
7-Quinolinol 99%
7-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 239…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00016730
CAS Number: 580-20-1
EINECS: 209-457-2
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Beige to light brown Powder
Infrared spectrum Conforms
Melting point 239 to 244 °C (Dec)
Titration with HClO4 ≥98.5 %