Quinaldine
Supplier: Merck
|
Warning
|
Synonyms:
2-Methylquinoline
22550-1L-F
22550-250ML-F
SIAL22550-1L-FEA
225
EUR
SIAL22550-1L-F
SIAL22550-250ML-F
Quinaldine
Quinaldine
Formula:
C₁₀H₉N MW: 143.19 g/mol Boiling Pt: 247.6 °C (1013 hPa) Melting Pt: –2 °C Density: 1.0585 g/cm³ (20 °C) Flash Pt: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006756 CAS Number: 91-63-4 EINECS: 202-085-1 Merck Index: 12,08228 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...