Qualitative filter papers, wet strengthened grades, grade 1573 and 1573 ½, Whatman™
Supplier: Whatman products (Cytiva)
10314712 10314714 10314744 10314745 10314747 10314751 10314752 10314753 10314766 10314765 10314726
515-6005EA 54.5 EUR
515-6005 515-6006 515-7004 515-7005 515-7006 515-7007 515-7008 515-7009 512-0645 512-0644 512-0610
Qualitative filter papers, wet strengthened grades, grade 1573 and 1573 ½, Whatman™
Filters Filter Papers Qualitative Filter Paper
A fast filter paper with high wet strength. It has a very smooth surface making it easy to scrape or wash off precipitate.
  • Folded filters for easy use in applications involving a conical filter funnel
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, and more.
Order Now
SPECIFICATIONS
Grade 1573 1573 ½
Environmentally Preferable
Weight 88 g/m² 88 g/m²
Thickness 170 µm 170 µm
Particle retention 25 µm 25 µm
Filtration speed 25 s (Herzberg) 25 s (Herzberg)