Dimethyl-2-aminoterephthalate 99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate
194780250 194780500
ACRO194780250EA 52.4 EUR
ACRO194780250 ACRO194780500
Dimethyl-2-aminoterephthalate 99%
Dimethyl 2-aminoterephthalate
Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00008427
CAS Number: 5372-81-6
EINECS: 226-364-2
Order Now
Specification Test Results
Appearance Yellow to beige to light green to brown Powder or crystals
Infrared spectrum Conforms
Melting point 131 to 134 °C
GC ≥98.5 %