Lanthanum(III) trifluoromethanesulphonate 99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Lanthanum(III) trifluoromethanesulphonate, Lanthanum trifluoromethanesulphonate, Lanthanum triflate
343910010 343910050
ACRO343910010EA 30.7 EUR
ACRO343910010 ACRO343910050
Lanthanum(III) trifluoromethanesulphonate 99%
Lanthanum (III) trifluoromethanesulphonate
Formula: C₃F₉LaO₉S₃
MW: 586.12 g/mol
Density: 1.7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00077560
CAS Number: 52093-26-2
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance White to light beige Powder or crystals
Infrared spectrum Conforms
Elemental Analysis 23.35 to 24.05 % (La)