3-Chlorophthalic anhydride 95+%
Supplier: Thermo Scientific
Warning
Synonyms: 4-Chloroisobenzofuran-1,3-dione
381290010 381290050 381290250
ACRO381290010EA 32.5 EUR
ACRO381290010 ACRO381290050 ACRO381290250
3-Chlorophthalic anhydride 95+%
3-Chlorophthalic anhydride
Formula: C₈H₃ClO₃
MW: 182.56 g/mol
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00023107
CAS Number: 117-21-5
EINECS: 204-179-8
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder or flakes and/or chunks
Infrared spectrum Conforms
Melting point 122 to 126 °C
Titration by coupling ≥95.0 %