1-Formylpiperazine 90%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 1-Piperazinecarboxaldehyde, 1-Piperazinecarbaldehyde
388490250 388491000
ACRO388490250EA 60.4 EUR
ACRO388490250 ACRO388491000
1-Formylpiperazine 90%
1-Formylpiperazine
Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
Boiling Pt: 105…107 °C (1 mmHg)
Density: 1.1 g/cm³
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005963
CAS Number: 7755-92-2
EINECS: 231-813-0
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5090 to 1.5130 (20°C, 589 nm)
GC ≥88 %