Magnesium trifluoromethanesulphonate
Supplier: Thermo Scientific
Danger
Synonyms: Magnesium trifluoromethanesulphonate, Magnesium triflate
22340.09 22340.18
22340.09EA 67.6 EUR
22340.09 22340.18
Magnesium trifluoromethanesulphonate
Magnesium trifluoromethanesulphonate
CAS No.: 60871-83-2 Molecular Formula: C2F6MgO6S2 Formula Weight: 322.43 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 310° Beilstein Reference: 705578 UN No.: 1759 MDL No.: MFCD00042601
Formula: C₂F₆MgO₆S₂
MW: 322.45 g/mol
Melting Pt: >310 °C
Storage Temperature: Ambient
MDL Number: MFCD00042601
CAS Number: 60871-83-2
ADR: 8,III