Triethyloxonium tetrafluoroborate 1.0 M in dichloromethane
Supplier: Thermo Scientific
Danger
A11678.AC A11678.AE A11678.AK A11678.AP A11678.AU
A11678.ACEA 44.4 EUR
A11678.AC A11678.AE A11678.AK A11678.AP A11678.AU
Triethyloxonium tetrafluoroborate 1.0 M in dichloromethane
Triethyloxonium tetrafluoroborate
CAS No.: 368-39-8 Molecular Formula: C6H15BF4O Formula Weight: 189.99 Storage Temperature: -30°C to -10°C Odour: Chloroform-like Beilstein Reference: 3598090 UN No.: 2922 MDL No.: MFCD00044423
Formula: C₆H₁₅BF₄O
MW: 189.99 g/mol
Density: 1.328
Storage Temperature: Freezer
MDL Number: MFCD00044423
CAS Number: 368-39-8
EINECS: 206-705-1
UN: 2922
ADR: 8,II