Methyl-4-(trifluoromethyl)benzoate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A15400.06
A15400.14
A15400.22
A15400.06EA
53
EUR
A15400.06
A15400.14
A15400.22
Methyl-4-(trifluoromethyl)benzoate ≥98%
Methyl 4-(trifluoromethyl)benzoate
Purity: 98%
CAS No.: 2967-66-0
Molecular Formula: C9H7F3O2
Formula Weight: 204.15
Flash Point: 82°(179°F)
Refractive Index: 1.4520
Melting Point: 13-14°
Boiling Point: 86-87°/10mm
Beilstein Reference: 1963288
MDL No.: MFCD00042324
Formula:
C₉H₇F₃O₂ Boiling Pt: 86…87 °C (10 mmHg) Melting Pt: 13…14 °C Density: 1.268 Flash Pt: 82 °C (179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042324 CAS Number: 2967-66-0 |