Methyl-2-bromo-5-chlorobenzoate ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: Methyl 2-bromo-5-chlorobenzoate
B25309.14 B25309.22
B25309.14EA 83.6 EUR
B25309.14 B25309.22
Methyl-2-bromo-5-chlorobenzoate ≥98%
Methyl 2-bromo-5-chlorobenzoate
Purity: 98% CAS No.: 27007-53-0 Molecular Formula: C8H6BrClO2 Formula Weight: 249.49 Melting Point: 37-40° MDL No.: MFCD00144763
Formula: C₈H₆BrClO₂
MW: 249.49 g/mol
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00144763
CAS Number: 27007-53-0