Methyl-3,5-dichlorobenzoate
Supplier: Thermo Scientific
Synonyms: Methyl 3,5-dichlorobenzoate
KM02512DA KM02512EA
MAYBKM02512DAEA 75.1 EUR
MAYBKM02512DA MAYBKM02512EA
Methyl-3,5-dichlorobenzoate
Methyl 3,5-dichlorobenzoate
Formula: C₈H₆Cl₂O₂
MW: 205.04 g/mol
MDL Number: MFCD00032710
CAS Number: 2905-67-1