Dimethyl-2-aminoterephthalate

Supplier: Thermo Scientific

Synonyms: Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate

SB01058.1 SB01058.10 SB01058.50
MAYBSB01058.1EA 0 EUR
MAYBSB01058.1 MAYBSB01058.10 MAYBSB01058.50
Dimethyl-2-aminoterephthalate
Dimethyl 2-aminoterephthalate
Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00008427
CAS Number: 5372-81-6
EINECS: 226-364-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR