Methyl 5-nitro-2-furoate

Supplier: Thermo Scientific

SB01301.1 SB01301.10 SB01301.50
MAYBSB01301.1EA 0 EUR
MAYBSB01301.1 MAYBSB01301.10 MAYBSB01301.50
Methyl 5-nitro-2-furoate
Methyl 5-nitro-2-furoate
Formula: C₆H₅NO₅
MW: 171.11 g/mol
MDL Number: MFCD00051956
CAS Number: 1874-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR