3-Picolyl chloride hydrochloride

Supplier: Thermo Scientific

Synonyms: 3-(Chloromethyl)-pyridinium chloride, 3-(Chloromethyl)pyridine hydrochloride

SB01796.1 SB01796.10 SB01796.50
MAYBSB01796.1EA 0 EUR
MAYBSB01796.1 MAYBSB01796.10 MAYBSB01796.50
3-Picolyl chloride hydrochloride
3-Picolyl chloride hydrochloride
Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
MDL Number: MFCD00012818
CAS Number: 6959-48-4
EINECS: 230-150-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR