2,3-Dichloro-6-nitrobenzonitrile
Supplier: Thermo Scientific
SEW05399.1
SEW05399.10
MAYBSEW05399.1EA
0
EUR
MAYBSEW05399.1
MAYBSEW05399.10
2,3-Dichloro-6-nitrobenzonitrile
2,3-Dichloro-6-nitrobenzonitrile
Formula:
C₇H₂Cl₂N₂O₂ |
MDL Number:
MFCD00728596 CAS Number: 2112-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...