Methyl-2-(trifluoromethyl)benzoate

Supplier: Thermo Scientific

Synonyms: Methyl 2-(trifluoromethyl)benzoate

TL00778.1 TL00778.10 TL00778.50
MAYBTL00778.1EA 0 EUR
MAYBTL00778.1 MAYBTL00778.10 MAYBTL00778.50
Methyl-2-(trifluoromethyl)benzoate
Methyl 2-(trifluoromethyl)benzoate
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
MDL Number: MFCD00236050
CAS Number: 344-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR