Brij® 35, powder

Supplier: Molekula

Synonyms: Brij® L23

MOLE28156962-1KGEA 206 EUR
MOLE28156962-1KG
Brij® 35, powder
Brij® 35
Formula: C₅₈H₁₁₈O₂₄
MW: 1199,56 g/mol
MDL Number: MFCD00080891
CAS Number: 9002-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR