Cadmium(II) cyclohexanebutyrate

Supplier: LGC Standards PROMOCHEM

PROONIST-1053AEA 838 EUR
PROONIST-1053A
Cadmium(II) cyclohexanebutyrate
Cadmium(II) cyclohexanebutyrate
Formula: C₂₀H₃₄CdO₄
MW: 450,9 g/mol
MDL Number: MFCD00036398
UN: 2570
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR