Bis(diphenylphosphino)methane, Sigma-Aldrich®

Supplier: Merck

Synonyms: [(1,1-Diphenylphosphino)methyl](diphenyl)phosphine, DPPM

M-Clarity™ quality level = MQ100

127566-5G
SIAL127566-5GEA 58.4 EUR
SIAL127566-5G
Bis(diphenylphosphino)methane, Sigma-Aldrich®
Bis(diphenylphosphino)methane
Formula: C₂₅H₂₂P₂
MW: 384.4 g/mol
MDL Number: MFCD00003537
CAS Number: 2071-20-7
EINECS: 218-194-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR