1,1'-Bis(diphenylphosphino)ferrocene, Sigma-Aldrich®
Supplier: Merck
Synonyms: DPPF, 1,1-Bis(diphenylphosphino)ferrocene
M-Clarity™ quality level = MQ200
177261-10G 177261-1G 177261-25G
SIAL177261-10GEA 200 EUR
SIAL177261-10G SIAL177261-1G SIAL177261-25G
1,1'-Bis(diphenylphosphino)ferrocene, Sigma-Aldrich®
1,1'-Bis(diphenylphosphino)ferrocene
Formula: C₃₄H₂₈FeP₂
MW: 554.39 g/mol
MDL Number: MFCD00001422
CAS Number: 12150-46-8
EINECS: 430-420-3