(-)-Menthone, Sigma-Aldrich®

Supplier: Merck

Synonyms: trans-Menthone, L-Menthone, (2S,5R)-2-Isopropyl-5-methylcyclohexanone, (1R,4S)-p-Menthan-3-one

M-Clarity™ quality level = MQ100

218235-100G 218235-25G
SIAL218235-100GEA 134 EUR
SIAL218235-100G SIAL218235-25G
(-)-Menthone, Sigma-Aldrich®
(-)-Menthone
Formula: C₁₀H₁₈O
MDL Number: MFCD00001634
CAS Number: 14073-97-3
EINECS: 237-926-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR