Silver perchlorate hydrate ≥99%, Sigma-Aldrich®
Supplier: Merck
226548-25G
226548-5G
SIAL226548-25GEA
130
EUR
SIAL226548-25G
SIAL226548-5G
Silver perchlorate hydrate ≥99%, Sigma-Aldrich®
Silver perchlorate hydrate
Formula:
AgClO₄·xH₂O MW: 207.32 g/mol (anhy) |
MDL Number:
MFCD00149128 CAS Number: 331717-44-3 EINECS: 232-035-4 UN: 1481 ADR: 5.1,II Merck Index: 12,08667 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...