Brij® L4 average Mn ~362, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Brij 30, Laureth-4, Brij L4, Polyoxyethylene (4) Lauryl Ether

M-Clarity™ quality level = MQ200

235989-100ML 235989-1L 235989-5ML
SIAL235989-100MLEA 26.3 EUR
SIAL235989-100ML SIAL235989-1L SIAL235989-5ML
Brij® L4 average Mn ~362, Sigma-Aldrich®
Brij® L4
Formula: (C₂₀H₄₂O₅)n
MDL Number: MFCD00043063
CAS Number: 9002-92-0
EINECS: 500-002-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR