Dipropyl phthalate, Sigma-Aldrich®

Supplier: Merck

290602-10G 290602-50G
SIAL290602-10GEA 52 EUR
SIAL290602-10G SIAL290602-50G
Dipropyl phthalate, Sigma-Aldrich®
Dipropyl phthalate
Formula: C₁₄H₁₈O₄
MW: 250.29 g/mol
MDL Number: MFCD00009371
CAS Number: 131-16-8
EINECS: 205-015-8
UN: 3082
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR