1-Acetylpiperazine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
359513-25G
359513-5G
SIAL359513-25GEA
141
EUR
SIAL359513-25G
SIAL359513-5G
1-Acetylpiperazine, Sigma-Aldrich®
1-Acetylpiperazine
Formula:
C₆H₁₂N₂O MW: 128.17 g/mol |
MDL Number:
MFCD00058676 CAS Number: 13889-98-0 EINECS: 237-659-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...