Sodium perchlorate hydrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
381225-100G 381225-500G
SIAL381225-100GEA 92.1 EUR
SIAL381225-100G SIAL381225-500G
Sodium perchlorate hydrate, Sigma-Aldrich®
Sodium perchlorate hydrate
Formula: ClNaO₄·H₂O
Storage Temperature: Ambient
CAS Number: 207683-20-3
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II
Merck Index: 12,08798