Samarium(III) trifluoromethanesulphonate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Samarium(III) trifluoromethanesulphonate

425710-10G 425710-1G
SIAL425710-10GEA 99.7 EUR
SIAL425710-10G SIAL425710-1G
Samarium(III) trifluoromethanesulphonate, Sigma-Aldrich®
Samarium (III) trifluoromethanesulphonate
Formula: C₃F₉O₉S₃Sm
MW: 597.57 g/mol
MDL Number: MFCD00192431
CAS Number: 52093-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR