Lithium trifluoromethanesulphonate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

481548-25G 481548-5G
SIAL481548-25GEA 153 EUR
SIAL481548-25G SIAL481548-5G
Lithium trifluoromethanesulphonate, Sigma-Aldrich®
Lithium trifluoromethanesulphonate
Formula: CF₃LiO₃S
MDL Number: MFCD00013227
CAS Number: 33454-82-9
EINECS: 251-528-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR