2-Phenoxyethanol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
M-Clarity™ quality level = MQ200
484318-18L 484318-1L
SIAL484318-18LEA 592 EUR
SIAL484318-18L SIAL484318-1L
2-Phenoxyethanol, Sigma-Aldrich®
2-Phenoxyethanol
Formula: C₆H₅OCH₂CH₂OH
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 13 °C
Density: 1.1094 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002857
CAS Number: 122-99-6
EINECS: 204-589-7
Merck Index: 12,07410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27cc4d39b806528798164961c23a1285d41d6a9&vid=a46b721cc69101d0c50dc49d9687445b395a3766&prid=web&clid=SAPDAM