2-Picolyl bromide hydrobromide, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-(Bromomethyl)pyridine hydrobromide

M-Clarity™ quality level = MQ100

491047-25G 491047-5G
SIAL491047-25GEA 125 EUR
SIAL491047-25G SIAL491047-5G
2-Picolyl bromide hydrobromide, Sigma-Aldrich®
2-Picolyl bromide hydrobromide
Formula: C₆H₇Br₂N
MW: 252.94 g/mol
MDL Number: MFCD01863544
CAS Number: 31106-82-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR