4-Picolyl bromide hydrobromide, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-(Bromomethyl)pyridine hydrobromide

M-Clarity™ quality level = MQ100

491748-25G 491748-5G
SIAL491748-25GEA 217 EUR
SIAL491748-25G SIAL491748-5G
4-Picolyl bromide hydrobromide, Sigma-Aldrich®
4-Picolyl bromide hydrobromide
Formula: C₆H₇Br₂N
MW: 252.94 g/mol
MDL Number: MFCD01863545
CAS Number: 73870-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR